GeneBLAzer™ ADRB2-CRE-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ ADRB2-CRE-bla CHO-K1 Cells

The GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells contain the human Adrenergic Beta-2 Receptor (ADRB2), (Accession No. NM_000024.3) stably integrated into the CellSensor™Read more
Have Questions?
Catalog number K1472
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ ADRB2-CRE-bla CHO-K1 Cells
Catalog numberK1472
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart

The GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells contain the human Adrenergic Beta-2 Receptor (ADRB2), (Accession No. NM_000024.3) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE).

The ADRB2-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Isoproterenol. In addition, ADRB2-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000024.3
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
Gene SymbolADRB2
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryADRB2
TypeCHO-K1 Cells
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells
• GeneBLAzer™ ADRB2-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.