GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 Cells

The GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells contain the human Complement Component 5a Receptor 1 (ADRA1B) stably integrated into the GeneBLAzer™ NFAT-blaRead more
Have Questions?
Catalog number K1471
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 Cells
Catalog numberK1471
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart

The GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells contain the human Complement Component 5a Receptor 1 (ADRA1B) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line. GeneBLAzer™ NFAT-bla CHO-K1 (Cat. No. K1447) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element stably integrated into CHO-K1 cells.

The ADRA1B-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using established ligands. In addition, ADRA1B-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000679.3
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
Gene SymbolADRA1B
Population StatusPool of Stable Cells
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryADRA1B
TypeCHO-K1 Cells
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells
• GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.