The GeneBLAzer™ ADRA1A-NFAT-bla CHO-K1 cells contain the human Adrenergic Alpha-1A Receptor (ADRA1A), (Accession No. NM_000680) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat No. K1078) contain a beta-lactamase (bla) reporter gene under control of the NFAT response element.
Both ADRA1A-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Phenylephrine. In addition, ADRA1A-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.