GeneBLAzer™ CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells contain the human Calcitonin Gene Related Peptide Receptor (CALCRL:RAMP1), (Accession No. NM_005795.3 NM_005855.2) stably integrated into the CellSensor™ CRE-bla Freestyle™ 293F cell line. CellSensor™ CRE-bla Freestyle™ 293F cells (Cat. No. K1130) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE).
CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells are functionally validated for Z'-factor and EC50 concentrations of Alpha-CGRP. In addition, CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.