Specifications
Alternate AgonistsL165, 041, GW 1929, GW 7647, GW 0742
AntagonistsT0070907, GW 9662, GW 6471
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene ID (Entrez)2101
Gene SymbolERRα
Primary AgonistRosiglitazone
Product LineGeneBLAzer™
Quantity1 plate
Reporter GeneBLA (Beta-Lactamase)
Target EntryESRRA, NR3B1, ERR1, ERR alpha
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetNuclear Receptors
Unit Size1 plate