GeneBLAzer™ LXR beta DA Assay Kit
Product Image
Thermo Scientific™

GeneBLAzer™ LXR beta DA Assay Kit

GeneBLAzer™LXR beta DA (Division Arrested) cells and LXR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the humanRead more
Have Questions?
Catalog number K1415
Price (USD)/ 1 plate
1,522.00
-
Add to cart
Price (USD)/ 1 plate
1,522.00
Add to cart
GeneBLAzer™ LXR beta DA Assay Kit
Catalog numberK1415
Price (USD)/ 1 plate
1,522.00
-
Add to cart

GeneBLAzer™LXR beta DA (Division Arrested) cells and LXR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-beta (LXR beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-LXR beta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both LXR beta DA cells and LXR beta-UAS-bla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of TO901317. In addition, LXR beta-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Alternate AgonistsDHT
AntagonistsCyp Acetate, Spirolactone, OHF=
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene ID (Entrez)7376
Gene SymbolLXRβ
Primary AgonistR1881
Product LineGeneBLAzer™
Quantity1 plate
Reporter GeneBLA (Beta-Lactamase)
Target EntryLXR beta, NR1H2, LXRB
TypeDivision-Arrested Cells Assay Kit
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetNuclear Receptors
Unit Size1 plate
Contents & Storage
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• LXR beta HEK 293T DA cells (K1415A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)