GeneBLAzer™ MR DA Assay Kit
Product Image
Thermo Scientific™

GeneBLAzer™ MR DA Assay Kit

GeneBLAzer™MR DA(Division Arrested) cells and MR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Mineralcorticoid Receptor (MR)Read more
Have Questions?
Catalog number K1409
Price (USD)/ 1 plate
1,540.00
-
Add to cart
Price (USD)/ 1 plate
1,540.00
Add to cart
GeneBLAzer™ MR DA Assay Kit
Catalog numberK1409
Price (USD)/ 1 plate
1,540.00
-
Add to cart

GeneBLAzer™MR DA(Division Arrested) cells and MR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Mineralcorticoid Receptor (MR) fused to the NAbinding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-MR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384- well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both MR DA cells and MRUAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of aldosterone. In addition, MR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and substrate loading time. Additional testing data using alternate stimuli are also available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Alternate AgonistsAM 580, Fenretinide, 9-cis- retinoic acid, Ch 55, TTNPB
AntagonistsLE 135
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene SymbolMR
Primary AgonistAll-trans-retinoic Acid
Product LineGeneBLAzer™
Quantity1 plate
Reporter GeneBLA (Beta-Lactamase)
Target EntryNR3C2, MR
TypeDivision-Arrested Cells Assay Kit
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetNuclear Receptors
Unit Size1 plate
Contents & Storage
Contains sufficient division-arrested cells and substrate to assay one 384-well plate
• MR HEK 293T DA cells (K1409A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)