GeneBLAzer™ PPAR delta DA Assay Kit
Product Image
Thermo Scientific™

GeneBLAzer™ PPAR delta DA Assay Kit

GeneBLAzer™ PPAR delta DA (Division Arrested) cells and PPAR delta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of theRead more
Have Questions?
Catalog number K1395
Price (USD)/ 1 plate
1,540.00
-
Add to cart
Price (USD)/ 1 plate
1,540.00
Add to cart
GeneBLAzer™ PPAR delta DA Assay Kit
Catalog numberK1395
Price (USD)/ 1 plate
1,540.00
-
Add to cart

GeneBLAzer™ PPAR delta DA (Division Arrested) cells and PPAR delta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-delta (PPAR delta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-PPAR delta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both PPAR delta DA cells and PPAR delta-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of L-165041. In addition, PPAR delta-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Alternate AgonistsEstrone, Genistein, Diethylstibestrol, Ethynylestradiol
Antagonists4-OH-tam+0.2 nM E2, Tam+0.2 nM E2, 4-OH-tamoxifen, Tamoxifen, RU486+0.2 nM E2, RU486
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene ID (Entrez)5467
Gene SymbolPPARδ
Primary Agonist17-β-estradiol
Product LineGeneBLAzer™
Quantity1 plate
Reporter GeneBLA (Beta-Lactamase)
Target EntryPPAR delta, NR1C2, PPARD
TypeDivision-Arrested Cells Assay Kit
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetNuclear Receptors
Unit Size1 plate
Contents & Storage
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• PPAR delta HEK 293T DA cells (K1395A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)