NovaFluor™ Antibody Conjugation Kits
NovaFluor™ Antibody Conjugation Kits
Invitrogen™

NovaFluor™ Antibody Conjugation Kits

Get the most out of your panel design by conjugating antibodies to NovaFluors: dyes specifically designed for spectral hygiene to have narrow emission spectra and minimal cross-laser excitation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeLabel or DyeEmission
K06T04L032Conjugation KitNovaFluor™ Violet 690765 nm
K06T04L008Conjugation KitNovaFluor™ Blue 510510 nm
K06T04L002Conjugation KitNovaFluor™ Blue 530530 nm
K06T04L009Conjugation KitNovaFluor™ Blue 555555 nm
K06T04L010Conjugation KitNovaFluor™ Blue 585585 nm
K06T04L003Conjugation KitNovaFluor™ Blue 610/30S610 nm
K06T04L011Conjugation KitNovaFluor™ Blue 610/70S610 nm
K06T04L004Conjugation KitNovaFluor™ Blue 660/40S660 nm
K06T04L021Conjugation KitNovaFluor™ Blue 690690 nm
K06T04L012Conjugation KitNovaFluor™ Blue 660/120S660 nm
K06T04L025Conjugation KitNovaFluor™ Blue 725725 nm
K06T04L026Conjugation KitNovaFluor™ Blue 760760 nm
K06T04L017Conjugation KitNovaFluor™ Red 660660 nm
K06T04L018Conjugation KitNovaFluor™ Red 685685 nm
K06T04L019Conjugation KitNovaFluor™ Red 700700 nm
K06T04L020Conjugation KitNovaFluro™ Red 710710 nm
K06T04L023Conjugation KitNovaFluor™ Red 725725 nm
K06T04L024Conjugation KitNovaFluor™ Red 755755 nm
K06T04L005Conjugation KitNovaFluor™ Yellow 570570 nm
K06T04L013Conjugation KitNovaFluor™ Yellow 590590 nm
K06T04L006Conjugation KitNovaFluor™ Yellow 610610 nm
K06T04L007Conjugation KitNovaFluor™ Yellow 660660 nm
K06T04L014Conjugation KitNovaFluor™ Yellow 690690 nm
K06T04L015Conjugation KitNovaFluor™ Yellow 700700 nm
K06T04L016Conjugation KitNovaFluor™ Yellow 730730 nm
K06T04L022Conjugation KitNovaFluor™ Yellow 755755 nm
K06T04L027Conjugation KitNovaFluor™ UV 765800 nm
K06T04L028Conjugation KitNovaFluor™ Violet 800810 nm
K06T04L029Conjugation KitNovaFluor™ Violet 745800 nm
K06T04L030Conjugation KitNovaFluor™ Yellow 810690 nm
K06T04L031Conjugation KitNovaFluor™ Blue 725745 nm
K06T04L033Conjugation KitNovaFluor™ Violet 690725 nm
Catalog number K06T04L032
Price (USD)/ 100 μg
500.00
-
Add to cart
Product Type:
Conjugation Kit
Label or Dye:
NovaFluor™ Violet 690
Emission:
765 nm
Price (USD)/ 100 μg
500.00
Add to cart
NovaFluor™ Antibody Conjugation Kits
Catalog numberK06T04L032
Price (USD)/ 100 μg
500.00
-
Add to cart
NovaFluor Conjugation Kits provide a simple workflow to conjugate a blue, yellow-green or red excitable fluorophore to your antibody of choice in just 3 easy steps. The fluorescence-by-design Phiton platform enables the addition of parameters to your current panels in discovery research and resolves stained populations with near-zero cross excitation, with a lack of emission in the violet channels that plague other fluorophores.

NovaFluor Conjugation Kit provides features including:

  • A clean and bright antibody label to increase the number of parameters on conventional and spectral flow cytometers
  • Resolve populations with limited/near-zero cross excitation and the lowest spread performance of any blue, yellow-green or red excited fluorescent labels
  • Simple 3-step protocol, with 60 sec. of hands-on time for conjugating 10–100 μg of antibody per label
  • Up to 70% less excitation when compared to PE tandems, resulting in decreased spillover and higher resolution

3-step conjugation method

  • Attach NovaFluor Linker to antibody of choice
  • Remove excess NovaFluor Linker from DNA-conjugate antibody
  • Anneal NovaFluor to oligo-conjugated antibody
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 μg
Product TypeConjugation Kit
Chemical ReactivityAmine
Label or DyeNovaFluor™ Violet 690
Label TypeOther Labels or Dyes
Product LineNovaFluor™
Emission765 nm
Excitation358 nm
Labeling TargetAntibody
Unit Size100 μg