Catalog Number | Product Type | Label or Dye | Emission |
---|---|---|---|
K06T04L002 | Conjugation Kit | NovaFluor™ Blue 530 | 530 nm |
K06T04L008 | Conjugation Kit | NovaFluor™ Blue 510 | 510 nm |
K06T04L009 | Conjugation Kit | NovaFluor™ Blue 555 | 555 nm |
K06T04L010 | Conjugation Kit | NovaFluor™ Blue 585 | 585 nm |
K06T04L003 | Conjugation Kit | NovaFluor™ Blue 610/30S | 610 nm |
K06T04L011 | Conjugation Kit | NovaFluor™ Blue 610/70S | 610 nm |
K06T04L004 | Conjugation Kit | NovaFluor™ Blue 660/40S | 660 nm |
K06T04L021 | Conjugation Kit | NovaFluor™ Blue 690 | 690 nm |
K06T04L012 | Conjugation Kit | NovaFluor™ Blue 660/120S | 660 nm |
K06T04L025 | Conjugation Kit | NovaFluor™ Blue 725 | 725 nm |
K06T04L026 | Conjugation Kit | NovaFluor™ Blue 760 | 760 nm |
K06T04L017 | Conjugation Kit | NovaFluor™ Red 660 | 660 nm |
K06T04L018 | Conjugation Kit | NovaFluor™ Red 685 | 685 nm |
K06T04L019 | Conjugation Kit | NovaFluor™ Red 700 | 700 nm |
K06T04L020 | Conjugation Kit | NovaFluro™ Red 710 | 710 nm |
K06T04L023 | Conjugation Kit | NovaFluor™ Red 725 | 725 nm |
K06T04L024 | Conjugation Kit | NovaFluor™ Red 755 | 755 nm |
K06T04L005 | Conjugation Kit | NovaFluor™ Yellow 570 | 570 nm |
K06T04L013 | Conjugation Kit | NovaFluor™ Yellow 590 | 590 nm |
K06T04L006 | Conjugation Kit | NovaFluor™ Yellow 610 | 610 nm |
K06T04L007 | Conjugation Kit | NovaFluor™ Yellow 660 | 660 nm |
K06T04L014 | Conjugation Kit | NovaFluor™ Yellow 690 | 690 nm |
K06T04L015 | Conjugation Kit | NovaFluor™ Yellow 700 | 700 nm |
K06T04L016 | Conjugation Kit | NovaFluor™ Yellow 730 | 730 nm |
K06T04L022 | Conjugation Kit | NovaFluor™ Yellow 755 | 755 nm |
K06T04L027 | Conjugation Kit | NovaFluor™ UV 765 | 800 nm |
K06T04L028 | Conjugation Kit | NovaFluor™ Violet 800 | 810 nm |
K06T04L029 | Conjugation Kit | NovaFluor™ Violet 745 | 800 nm |
K06T04L030 | Conjugation Kit | NovaFluor™ Yellow 810 | 690 nm |
K06T04L031 | Conjugation Kit | NovaFluor™ Blue 725 | 745 nm |
K06T04L032 | Conjugation Kit | NovaFluor™ Violet 690 | 765 nm |
K06T04L033 | Conjugation Kit | NovaFluor™ Violet 690 | 725 nm |
NovaFluor Conjugation Kit provides features including:
3-step conjugation method