Artemisinin, 98%, Thermo Scientific Chemicals
Artemisinin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Artemisinin, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65406.06
also known as J65406-06
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
Add to cart
Artemisinin, 98%, Thermo Scientific Chemicals
Catalog numberJ65406.06
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(1R,4S,5R,8S,9R,12S,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-one
Molecular FormulaC15H22O5
InChI KeyBLUAFEHZUWYNDE-NNWCWBAJSA-N
SMILESC[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4
Molecular Weight (g/mol)282.34
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Optical Rotation+70.0 to +80.0°
Appearance (Color)White
Infrared spectrumConforms
Assay from Supplier's CofA≥97.5% (HPLC)
Artemisinin inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Artemisinin crosses the blood-brain barrier and is an inhibitor of human NOS2 (iNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 100mM in DMSO and to 75mM in ethanol

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
RUO – Research Use Only

General References:

  1. Jurg Utzinger; Shu-Hua Xiao; Marcel Tanner; Jennifer Keiser. Artemisinins for schistosomiasis and beyond. Current Opinion in Investigational Drugs. 2007, 8, (2), 105-116.
  2. Rita Piedade; José Pedro Gil. The pharmacogenetics of antimalaria artemisinin combination therapy. Expert Opinion on Drug Metabolism and Toxicology. 2011, 7, (10), 1185-1200.