Anza™ T4 DNA Ligase Master Mix
Anza™ T4 DNA Ligase Master Mix
Invitrogen™

Anza™ T4 DNA Ligase Master Mix

The Invitrogen Anza™ T4 DNA Ligase Master Mix facilitates the joining of abutted 5’-phosphate and 3’-hydroxyl termini in duplex DNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
IVGN210450 Reactions
IVGN2108200 Reactions
Catalog number IVGN2104
Price (USD)/ 50 reactions
261.65
Online exclusive
278.00 
Save 16.35 (6%)
-
Add to cart
Quantity:
50 Reactions
Price (USD)/ 50 reactions
261.65
Online exclusive
278.00 
Save 16.35 (6%)
Add to cart
Anza™ T4 DNA Ligase Master Mix
Catalog numberIVGN2104
Price (USD)/ 50 reactions
261.65
Online exclusive
278.00 
Save 16.35 (6%)
-
Add to cart
The Invitrogen Anza™ T4 DNA Ligase Master Mix facilitates the joining of abutted 5’-phosphate and 3’-hydroxyl termini in duplex DNA through the formation of a phosphodiester bond. It can be used to join DNA fragments with both sticky ends and blunt ends, and to repair nicks in double-stranded DNA with 3'-hydroxyl and 5'-phosphate ends. Anza T4 DNA Ligase is formulated as a 4X concentrated master mix. Ligation can be performed with DNA in water, TE, elution buffer, or 1X Anza™ buffers.

Benefits:
• Ligation complete in 15 minutes at room temperature
• A single ligase master mix for both sticky-end and blunt-end ligation reactions
• Ready-to-use master mix format reduces pipetting steps
• 4X concentration enables more dilute DNA to be used in ligation reaction without concentrating

Anza T4 DNA Ligase Master Mix is part of the Anza™ Restriction Enzyme Cloning System, unifying the traditional cloning processes. Other Anza DNA Modifying Enzymes include the Anza T4 PNK Kit, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration4X
Shipping ConditionApproved for shipment on Wet or Dry Ice
EnzymeT4 DNA Ligase
Compatible BufferElution Buffer, 1X Anza Buffer
Quantity50 Reactions
Product TypeT4 DNA Ligase Master Mix
Product LineAnza™
Unit Size50 reactions
Contents & Storage
250 μL Anza T4 DNA Ligase Master Mix

Store at -5 to -30°C.