Anza™ Buffer Set
Product Image
Invitrogen™
Anza™ Buffer Set
The Invitrogen™ Anza™ Buffer Set includes both the Anza™ Buffer (10X) and the Anza™ Red Buffer (10X). The Anza buffersRead more
Have Questions?
Catalog number IVGN2008
Price (USD)/ 1 set
32.87
-
Add to cart
Price (USD)/ 1 set
32.87
Add to cart
Anza™ Buffer Set
Catalog numberIVGN2008
Price (USD)/ 1 set
32.87
-
Add to cart
The Invitrogen™ Anza™ Buffer Set includes both the Anza™ Buffer (10X) and the Anza™ Red Buffer (10X). The Anza buffers are proprietary buffer forumulations for use with the Anza™ Restriction Enzyme Cloning System. All Anza restriction enzymes digest DNA to completion in 15 minutes in the Anza buffers, and all Anza DNA modifying enzymes are fully functional in the Anza buffers.
Benefits:
• One buffer for all restriction enzymes—no buffer look-up
• Multiple digestions simplified with a single buffer
• Direct loading from digestion reaction onto gel with Anza Red Buffer
• Streamlined procedures with Anza DNA modifying enzymes
The Anza 10X Buffer Set is part of the Anza Restriction Enzyme Cloning System. For more information on the system, visit www.thermofisher.com/anza.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping ConditionApproved for shipment on Wet or Dry Ice
Quantity1 set
Product TypeBuffer Set
Research CategoryTraditional Cloning
Product LineAnza™
Unit Size1 set
Contents & Storage
2 mL Anza Buffer (10X)
2 mL Anza Red Buffer (10X)
Store at -5 to -30°C.