Anza™ 17 KpnI
Anza™ 17 KpnI
Invitrogen™
Anza™ 17 KpnI
5' G G T A C ↓ C 3'3' C ↑ C A T G G 5'Invitrogen™ Anza™ 17 KpnIRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
IVGN01763000 U
IVGN017815,000 U
Catalog number IVGN0176
Price (USD)/ 3000 units
104.00
-
Add to cart
Quantity:
3000 U
Price (USD)/ 3000 units
104.00
Add to cart
Anza™ 17 KpnI
Catalog numberIVGN0176
Price (USD)/ 3000 units
104.00
-
Add to cart
5'   G    G    T    A    C ↓ C   3'
3'   C ↑ C    A    T    G    G   5'
Invitrogen™ Anza™ 17 KpnI is a restriction enzyme that cuts DNA at this recognition site: GGTAC^C, completely digesting the DNA in 15 minutes at 37°C.
For superior convenience, a single buffer and protocol are used with all Anza™ restriction enzymes. The single Anza buffer allows multiple restriction enzymes to be used simultaneously in one reaction. This helps eliminate the need for sequential digests and the DNA clean-up necessary when changing buffers. Additionally, the Anza buffer and restriction enzymes have been designed for flexibility in digestion times. Digestions of up to 16 hours can be carried out without star activity.
Convenient—all Anza enzymes are 100% functional in the single Anza buffer
Fast—complete digestion in 15 minutes
Flexible—overnight digestion without star activity
Simple—a single digestion protocol for all DNA types
Complete system—incorporates Anza DNA Modifying Enzymes for a complete cloning experience
Anza DNA Modifying Enzymes include the Anza T4 DNA Ligase Master Mix, Anza Alkaline Phosphatase Kit, Anza T4 PNK Kit, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit. Use these enzymes for a complete cloning workflow.
Anza Red Buffer, which contains a density gradient and tracking dyes, is supplied for added convenience. Digestion reactions are as efficient in the Anza Red Buffer as in the Anza Buffer. Use of the Anza Red Buffer helps reduce pipetting steps when preparing multiple digestion reactions and a subsequent step includes agarose gel electrophoresis. For applications that require product analysis by fluorescence excitation, the colorless Anza Buffer without the tracking dyes is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping ConditionApproved for shipment on Wet or Dry Ice
Methylation SensitivityDcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
EnzymeKpn I
Compatible BufferAnza™ 10X Red Buffer, Anza™ 10X Buffer
Sensitive to Heat InactivationNo
Optimal Reaction Temperature37°C
Type IIS RENo
Quantity3000 U
Product TypeRestriction Enzyme Cloning System
Research CategoryTraditional Cloning
Product LineAnza™
Unit Size3000 units
Contents & Storage
300 μL Anza 17 KpnI
1 mL Anza Buffer (10X)
1 mL Anza Red Buffer (10X)
Store at -20°C.