Invivofectamine™ 3.0 Reagent
Invivofectamine™ 3.0 Reagent
Invitrogen™

Invivofectamine™ 3.0 Reagent

Invivofectamine™ 3.0 Reagent is an animal origin-free lipid nanoparticle designed for high efficiency in vivo delivery of siRNA and miRNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
IVF300110
IVF300550 Reactions
Catalog number IVF3001
Price (USD)/ 10 reactions
1,729.65
Online exclusive
1848.00 
Save 118.35 (6%)
-
Add to cart
No. of Reactions:
10
Price (USD)/ 10 reactions
1,729.65
Online exclusive
1848.00 
Save 118.35 (6%)
Add to cart
Invivofectamine™ 3.0 Reagent
Catalog numberIVF3001
Price (USD)/ 10 reactions
1,729.65
Online exclusive
1848.00 
Save 118.35 (6%)
-
Add to cart
Invivofectamine™ 3.0 Reagent is an animal origin-free lipid nanoparticle designed for high efficiency in vivo delivery of siRNA and miRNA to mouse liver cells following tail vein injection.

Using Invivofectamine™ 3.0 Reagent
Invivofectamine™ 3.0 Reagent is ideally suited for in vivo delivery of siRNA and miRNA duplexes. Invivofectamine™ 3.0/RNA complexes are delivered using a low-volume, low-pressure method with no toxicity and no stress response in the animal. Invivofectamine™ 3.0 can also be combined with BLOCK-iT™ fluorescent controls to track the biodistribution of the siRNA or miRNA.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free
Cell TypeHepatocytes
Sample TypeSynthetic siRNA, siRNA, miRNA, RNAi
Transfection TechniqueLipid-based Transfection, Lipid Nanoparticles, LNPs
SpeciesSmall Animal Models
For Use With (Application)Transfection
No. of Reactions10
Product LineInvivofectamine™
Product TypeReagent
RNAi TypesiRNA, miRNA
Serum CompatibleYes
Shipping ConditionDry Ice
Unit Size10 reactions