ARL™ EQUINOX 100 X-ray Diffractometer
ARL™ EQUINOX 100 X-ray Diffractometer
Thermo Scientific™
ARL™ EQUINOX 100 X-ray Diffractometer
Achieve high resolution analysis in minutes with a compact, transportable bench top X-Ray diffractometer for QA/QC, academic, and routine X-ray diffraction applications.
Have Questions?
Catalog number IQLAAHGABLFAQLMBIX
Price (USD)/ Each
-
Request A Quote
Description:
ARL EQUINOX 100 X-ray Diffractometer
Get a dedicated XRD instrument designed for QA/QC, academic, and routine X-ray diffraction applications with the compact, affordable Thermo Scientific™ ARL™ EQUINOX 100 X-ray Diffractometer. Occupying a third of the space of a conventional diffractometer, this stand alone, transportable benchtop instrument offers true flexibility. No external water cooling is required, and it plugs into a standard electrical power outlet. This instrument simplifies operation without sacrificing speed or performance. Thanks to a specific real time detection mode, measurements are completed in a few seconds.
  • Unique ARL EQUINOX curved detector measures all diffraction peaks simultaneously across a wide angular range. Analysis is completed in just a few minutes on most samples regardless of resolution requirements.
  • Measurement in real time over 110° 2Ø
  • No scanning required
  • Micro-focus X-Ray source with a high intensity optic
  • No external water cooling and plugging into a standard power supply
  • Many sample holders are available for: powders in transmission and reflection mode, bulks, thin films, in-situ studies, as well as automated analysis thanks to a dedicated sample changer
  • Easy to use and to install in various environment: laboratory, mobile laboratory, van
  • Treatment and applications: Phase identification, Quantification, Degree of crystallinity, Cell parameters, Crystallite size, Lattice strain, Crystal structure, Rietveld analysis, Transition phase, Thin film analysis (GIXRD, XRR)
  • Domains of application: Materials Science, Minerals, Geology, Nanomaterials, Chemicals, Pharmaceuticals, Energy materials, Catalysts, Ceramics, Polymers, Metallurgy, Semiconductors, Mining (cement, …), Teaching
Innovative solutions from exploration to mining production
From exploration to analysis and processing, either online or in the laboratory, we have everything necessary to meet the specifications of quality, environment and productivity. Thermo Scientific mining solutions provide reliable, accurate data and essential information for optimal process control. Learn more about these solution for mining production.
Specifications
TypeX-ray Diffractometer
DescriptionARL EQUINOX 100 X-ray Diffractometer
Unit SizeEach