Speed characterization of complex glycans; use SimGlycan™ software to interpret the rich MSn data produced by Thermo Scientific™ mass spectrometers.
Speed the characterization of complex glycans using SimGlycan™ software from PREMIER Biosoft. SimGlycan software processes and interprets the multi-stage/sequential (MSn) data produced by Thermo Scientific™ Mass Spectrometers, a capability essential to determining glycan heterogeneity and isomeric forms. The software automatically matches experimental mass spectra against a comprehensive database and generates a scored list of candidate structures. More relevant information about the proposed structure, including the glycan class, pathway and enzyme, is provided via interactive links.