OMNIC Paradigm Software
OMNIC Paradigm Software
Thermo Scientific™
OMNIC Paradigm Software
Automate workflows, build libraries and accelerate your spectral analysis using intuitive software that generates fast, reliable answers. This software complements the OMNIC Paradigm Touchscreen Software for the Nicolet Summit FTIR Spectrometer.
Have Questions?
Catalog number IQLAADGABZFAILMBKO
Price (USD)/ Each
-
Request A Quote
Description:
OMNIC Paradigm Software
For Use With (Application):
Material Characterization, Identification, Verification
Streamline material analysis with the Thermo Scientific™ OMNIC™ Paradigm Software. The user-friendly dashboard screen helps you view instrument status and recent work, process your spectra, conduct multi-component searching, and create new libraries. Designed with lab managers and science educators in mind, this software helps automate workflows using an intuitive drag-and-drop workflow creator. Work remotely and collaborate with colleagues around the globe when you upload OMNIC Paradigm data to the cloud using the Thermo Scientific OMNIC Anywhere Application.
Access the latest OMNIC Paradigm User Guide
Take your analysis further than our original OMNIC Software with the next generation OMNIC Paradigm Software. Process spectra, generate custom workflows, create libraries and more in an intuitive, modern software interface.
The OMNIC Paradigm Software offers:
  • Dashboard/Home screen for quick access of recent work
  • Visual, drag-and-drop workflow creator with example templates
  • Flexible, one-click library creation
  • Pre-defined reporting templates exportable to Microsoft Office Suite
  • Multi-component search functionality
  • Full spectral processing and analysis tools (baseline correction, spectra math, peak area, peak height, etc.)
  • Quantification prediction for Beer’s Law, PLS, CLS, etc.
  • Multiple project windows and multi-monitor support
Continue exploring OMNIC Paradigm Software at thermofisher.com/paradigm
Specifications
For Use With (Equipment)Nicolet Summit FTIR Spectrometer
DescriptionOMNIC Paradigm Software
For Use With (Application)Material Characterization, Identification, Verification
Unit SizeEach