Get significant increases in resolution and throughput in this high-pressure modular IC system for standard bore and microbore columns.
Catalog Number | Description |
---|---|
ICS6000-013 | Dionex ICS-6000 2D-IC Standard Bore-Microbore System |
ICS6000-007 | Dionex ICS-6000 Single Channel Standard Bore IC System |
ICS6000-008 | Dionex ICS-6000 Single Channel Gradient Standard Bore IC System |
ICS6000-009 | Dionex ICS-6000 Single Channel Standard Bore RFIC System |
ICS6000-010 | Dionex ICS-6000 Single Channel Microbore RFIC System |
ICS6000-011 | Dionex ICS-6000 Dual Channel Standard Bore RFIC System |
ICS6000-012 | Dionex ICS-6000 Dual Channel Microbore RFIC System |
ICS6000-014 | Dionex ICS-6000 Carbohydrate Analysis Standard Bore System |
ICS6000-015 | Dionex ICS-6000 BioIC System with ED for Carbohydrates (Standard Bore) and AAA-Direct (Microbore) System |
Get significant increases in resolution and throughput using the world's first modular ion chromatography (IC) system capable of operation up to 5000 psi. The Thermo Scientific™ Dionex™ ICS-6000 Standard Bore and Microbore System can operate continuously at up to 5000 psi when configured as a Reagent-Free™ IC (RFIC™) system. Get higher resolution using smaller particle size (4μm) columns, faster run times without sacrificing needed resolution using higher flow rates and 150mm columns, or higher resolution with standard flow rates and 250mm columns. The ability to configure this flexible, modular system for an unmatched variety of applications.
Broad variety of modules and related products
Flexible choice of detectors