Dionex™ ICS-6000 Standard Bore and Microbore HPIC™ Systems
Dionex™ ICS-6000 Standard Bore and Microbore HPIC™ Systems
Actual product may vary
Thermo Scientific™
Dionex™ ICS-6000 Standard Bore and Microbore HPIC™ Systems
Get significant increases in resolution and throughput in this high-pressure modular IC system for standard bore and microbore columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
ICS6000-007Dionex ICS-6000 Single Channel Standard Bore IC System
ICS6000-008Dionex ICS-6000 Single Channel Gradient Standard Bore IC System
ICS6000-009Dionex ICS-6000 Single Channel Standard Bore RFIC System
ICS6000-010Dionex ICS-6000 Single Channel Microbore RFIC System
ICS6000-011Dionex ICS-6000 Dual Channel Standard Bore RFIC System
ICS6000-012Dionex ICS-6000 Dual Channel Microbore RFIC System
ICS6000-013Dionex ICS-6000 2D-IC Standard Bore-Microbore System
ICS6000-014Dionex ICS-6000 Carbohydrate Analysis Standard Bore System
ICS6000-015Dionex ICS-6000 BioIC System with ED for Carbohydrates (Standard Bore) and AAA-Direct (Microbore) System
Catalog number ICS6000-007
Price (USD)/ Each
-
Request A Quote
Description:
Dionex ICS-6000 Single Channel Standard Bore IC System
Get significant increases in resolution and throughput using the world's first modular ion chromatography (IC) system capable of operation up to 5000 psi. The Thermo Scientific™ Dionex™ ICS-6000 Standard Bore and Microbore System can operate continuously at up to 5000 psi when configured as a Reagent-Free™ IC (RFIC™) system. Get higher resolution using smaller particle size (4μm) columns, faster run times without sacrificing needed resolution using higher flow rates and 150mm columns, or higher resolution with standard flow rates and 250mm columns. The ability to configure this flexible, modular system for an unmatched variety of applications.
Outstanding sensitivity plus convenience
  • Excellent sensitivity, stability, and ease of use
  • Enhanced baseline stability and sensitivity with top-of-the-line flow rate accuracy, eluent generator electronics stability, and conductivity cell temperature control
Optimized peak resolution with a choice of isocratic and electrolytic gradient separations
  • Flexible support for microbore (1–3mm i.d.) and standard bore (3–7mm i.d.) columns
  • Wide variety of Thermo Scientific standard and microbore columns
  • Compatible with New 4μm Columns
Separations twice as fast without compromising resolution
  • Significantly increased resolution from current methodologies at increased flow rates
  • Higher resolution and dramatically better performance using smaller particle size (4μm) columns
  • Faster run times with higher flow rates using 150mm columns, or higher resolution with standard flow rates using 250mm columns
Outstanding flexibility to configure the system you need
Broad variety of modules and related products
  • Dionex ICS-6000 standard (SP) and dual (DP) pumps for high-performance separations
  • Dionex ICS-6000 EG Eluent Generator for the benefits of an RFIC system
  • High-performance ICS-6000 DC Detector/Chromatography Compartment
  • IC and RFIC autosamplers and injectors for automation, from simple injections to complex sample preparation
  • Thermo Scientific™ Dionex Chromeleon™ Chromatography Data System software
Flexible choice of detectors
  • Dionex ICS-6000 CD Conductivity Detector in capillary and analytical formats
  • Dionex ICS-6000 ED Electrochemical Detector for high-sensitivity determination of difficult analytes
  • Dionex ICS-6000 Series VWD Variable Wavelength Detector and PDA Photodiode Array Detector for optical absorbance detection
  • Mass spectrometric detection for selective analysis
  • Inductively Coupled Plasma Mass Spectrometry (ICP-MS) for elemental speciation analysis
Specifications
TypeSingle Channel Standard Bore IC System
Includes1 x SP Pump, Isocratic with Degas<br /> 1 x DC (SB) with Dual Temperature Zone, One Injection Valve<br /> 1 x CD Detector (with Cell)<br /> 1 x EO Eluent Organizer Tray with two 2 Liter bottles<br /> 1 x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical)<br /> 1 x Consumable Device Monitor<br /> 1 x Tablet Kit (with arm, holder, and connectivity kit)
For Use With (Application)Ion Chromatography
DescriptionDionex ICS-6000 Single Channel Standard Bore IC System
Unit SizeEach