MAS™ Immunology Controls
MAS™ Immunology Controls
Actual product may vary
Thermo Scientific™
MAS™ Immunology Controls
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte immunology controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLevel
IC-303Immunology Controls, Assayed3
IC-101Immunology Controls, Assayed1
IC-202Immunology Controls, Assayed2
IC-V1Immunology Controls, Vista Vials, Assayed1
IC-V2Immunology Controls, Vista Vials, Assayed2
IC-V3Immunology Controls, Vista Vials, Assayed3
Catalog number IC-303
Price (USD)/ Each
-
Request A Quote
Description:
Immunology Controls, Assayed
Level:
3
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ Immunology Controls. These controls are also available in a convenient Vista™ vial format.
  • Assayed for most major instrument systems
  • Three levels provide monitoring of the full assay range
  • Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin and Rheumatoid Factor
  • Human-serum matrix
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionImmunology Controls, Assayed
Detectable AnalytesAlbumin, a-1-Acid Glycoprotein, a-1-Antitrypsin, a-2-Macroglobulin, Antistreptolysin, Antithrombin III, APO A (HDL Concentrate), APO B (LDL Concentrate), beta-2-microglobulin (B2M), C3 Complement, C4 Complement, Ceruloplasmin, Haptoglobin, IgA, IgE, IgG, IgM, Kappa Light Chain, Lambda Light Chain, Prealbumin, Properdin B, Rheumatoid Factor, Total Protein, Transferrin (TRF)
Sample TypeSerum
TypeControl
Level3
Packaging6 Pk.
Quantity3 mL
Unit SizeEach
Contents & Storage
Two year shelf life from date of manufacture at 2° to 8°C; 30 day open vial stability for all analytes at 2° to 8°C