Catalog Number | Product Type |
---|---|
I-APP00-00 | Sheet Extrusion Application Package |
I-APP00-01 | Cast Film Extrusion Application Package |
The IPlus! Measurement and Control System for Extrusion combines advanced sensor technology and powerful software in an intuitive, easy-to-use system to provide high performance and a low cost of ownership. The system incorporates advanced application packages for film and sheet manufacturing to increase the quality, consistency and performance characteristics of cast film and sheet products.
The suite of Thermo Scientific advanced machine-direction and cross-direction controls improve both quality and productivity, delivering consistent results and an impressive return on investment. The Machine Direction (MD) thickness control option is used to automatically maintain the extruded product to thickness specifications, while the Automatic Profile Control (APC) option controls auto dies to achieve cross-direction uniformity of extruded film and sheet.
Features:
Operator Interface:
Advanced Wonderware™ user interface with Windows operating system offers easier data management and archiving. Desktop interface and/or console configurations are available.
Control Options: