Human Plateable Hepatocytes, Induction Qualified
Human Plateable Hepatocytes, Induction Qualified
Gibco™

Human Plateable Hepatocytes, Induction Qualified

These plateable cryopreserved human hepatocytes are inducible and suitable for use in experiments monitoring in vitro enzyme induction. Cells perRead more
Have Questions?
Catalog number HMCPIS
Price (USD)/ 1.5 mL
1,978.00
-
Add to cart
Price (USD)/ 1.5 mL
1,978.00
Add to cart
Human Plateable Hepatocytes, Induction Qualified
Catalog numberHMCPIS
Price (USD)/ 1.5 mL
1,978.00
-
Add to cart
These plateable cryopreserved human hepatocytes are inducible and suitable for use in experiments monitoring in vitro enzyme induction.

Cells per vial: 4-8 million

The potentials of known clinically relevant enzyme inducers represented by Omeprazole (OMP, a prototypical CYP1A2 enzyme inducer, 100 μM), phenobarbital (PB, a prototypical CYP2B6 inducer, 1000 μM) and rifampicin (RIF, a prototypical CYP3A enzyme inducer, 10 μM) have been assessed to verify that each lot is inducible.

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, proper cell morphology, and high levels of induction increase the ability to draw in vitro/in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
• Viability ≥ 70%
• Attachment efficiency
• Monolayer formation and integrity
• Confluency >80% on Day 5
• Tested for plating in 96 well format
• Phase I enzyme activities
• Levels of CYP1A2, CYP2B6, and CYP3A4 induction
• Relevant CYP450 polymorphisms

Related Links:

Learn more about our cryopreserved hepatocytes
Learn about our full line of hepatocytes
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHepatocytes
Culture TypeMammalian Cell Culture
FormCryopreserved
SpeciesHuman
Donor SourceSingle Donor
No. of Cells4-8 x 10^6
Product LineGibco™
Product TypeHepatocytes
Quantity1 vial
Unit Size1.5 mL
Contents & Storage
Store in a cryostorage dewar or -135°C freezer.