Sensititre™ Gram Positive GPID ID Plate
Sensititre™ Gram Positive GPID ID Plate
Thermo Scientific™
Sensititre™ Gram Positive GPID ID Plate
Obtain species level identification of Gram positive organisms with overnight incubation using this convenient identification plate.
Have Questions?
Catalog number GPID
Price (USD)/ Each
-
Request A Quote
Description:
Sensititre Gram Positive Identification Plate
Quantity:
10/Pk.
Obtain species-level identification of Gram-positive organisms with overnight incubation using the Thermo Scientific™ Sensititre™ Gram Positive GPID ID Plate. If extended incubation is required for identification, the same plate can be re-incubated, eliminating unnecessary waste.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSensititre Gram Positive Identification Plate
Product TypeIdentification Test
Target Organism ClassGram-positive Organisms
Test TypePresumptive Identification
Isolates Per Plate3
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Unit SizeEach