Simplify testing of non-fastidious Gram-positive isolates with this plate containing CLSI breakpoint ranges for Ceftaroline and Telavancin
Perform susceptibility testing of non-fastidious Gram-positive isolates on a single platform with Thermo Scientific™ Sensititre™ Gram Positive GPALL3F AST Plate. The convenient standard AST plate includes updated CLSI breakpoints for Ceftaroline and Telavancin.
Achieve reliable, consistent results for non-fastidious Gram-positive isolates with the Sensititre Gram Positive GPALL3F AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.