E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 2%
Please note: Instrument packages that include E-Gels may encounter delivery delays.
E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 2%
Invitrogen™

E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 2%

The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, EX 2%, provides everything you need to quickly start using precastRead more
Have Questions?
Catalog number G8342ST
Price (USD)/ 1 kit
3,191.65
Online exclusive
3515.00 
Save 323.35 (9%)
-
Add to cart
Price (USD)/ 1 kit
3,191.65
Online exclusive
3515.00 
Save 323.35 (9%)
Add to cart
E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 2%
Catalog numberG8342ST
Price (USD)/ 1 kit
3,191.65
Online exclusive
3515.00 
Save 323.35 (9%)
-
Add to cart
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, EX 2%, provides everything you need to quickly start using precast E-Gel EX 2% agarose gels on the system for a streamlined gel electrophoresis and gel imaging workflow.

The kit includes:
• 10 E-Gel EX agarose gels, 2% (Cat. No. G401002)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
• Gel opening knife (Cat. No. EI9010)

Learn more about the E-Gel Power Snap Electrophoresis System ›

Learn more about precast E-Gel EX agarose gel technology ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeElectrophoresis System Starter Kit
Quantity1 kit
Shipping ConditionRoom Temperature
Max. Voltage240 V AC (Power Supply), 48 V DC (Electrophoresis Device)
Unit Size1 kit
Contents & Storage
• 1 electrophoresis device
• 1 camera device
• 10 E-Gel gels EX 2%
• 1 vial E-Gel 50 bp DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter
• 1 knife

Store at room temperature.