Catalog Number | Gel Percentage | Separation Range | Stain Type | Quantity |
---|---|---|---|---|
G820802 | 2% | 50 to 3000 bp | SYBR™ Safe | 8 Gels |
G800801 | 1% | 400 to 10,000 bp | Ethidium Bromide | 8 Gels |
G820801 | 1% | 400 to 10,000 bp | SYBR™ Safe | 8 Gels |
G820841 | 1% | 400 to 10,000 bp | SYBR™ Safe | 4 x 8 Gels |
G800802 | 2% | 50 to 3000 bp | Ethidium Bromide | 8 Gels |
G820842 | 2% | 50 to 3000 bp | SYBR™ Safe | 4 x 8 Gels |
G800804 | 4% | 10 to 400 bp | Ethidium Bromide | 8 Gels |
G820804 | 4% | 10 to 500 bp | SYBR™ Safe | 8 Gels |
G820844 | 4% | 10 to 500 bp | SYBR™ Safe | 4 x 8 Gels |
Each gel contains 48 sample wells and 4 marker wells in 1%, 2%, or 4% high-resolution agarose with a 3.2-cm run length. The various agarose percentages are used to separate DNA fragments of different sizes:
Each cassette is individually labeled with an EAN39 bar code that is recognized by most commercially available robotic bar code readers.
Features of E-Gel gels:
Less hazardous option
Choose gels with SYBR Safe DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide and UV. Precast E-Gel agarose gels with SYBR Safe stain, combined with a blue-light trans-illumination, offer better sample integrity and a less hazardous work environment. Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is less hazardous for the user, less hazardous for the environment, and can save institutional overhead costs for disposal.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng sample DNA per band and a separation time three times faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.
Running and imaging devices
High-throughput E-Gel agarose gels require the Invitrogen E-Base Electrophoresis Device for gel running.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with SYBR Safe stain are available in 1%, 2%, and 4% gel percentages.