E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX 1%
Please note: Instrument packages that include E-Gels may encounter delivery delays.
Invitrogen™
E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX 1%
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX 1%, provides everything you need to quickly start using precastRead more
Have Questions?
Catalog number G8141ST
Price (USD)/ 1 kit
1,908.65
Online exclusive
2015.00
Save 106.35 (5%)
-
Add to cart
Price (USD)/ 1 kit
1,908.65
Online exclusive
2015.00
Save 106.35 (5%)
Add to cart
E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX 1%
Catalog numberG8141ST
Price (USD)/ 1 kit
1,908.65
Online exclusive
2015.00
Save 106.35 (5%)
-
Add to cart
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX 1%, provides everything you need to quickly start using precast E-Gel EX 1% agarose gels on the device.
The kit includes: • 10 E-Gel EX agarose gels, 1% (Cat. No. G401001) • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100) • E-Gel 1 Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091) • Gel opening knife (Cat. No. EI9010)