Add a new dimension of selectivity for every proteomic laboratory with FAIMS Pro interface for HPLC mass spectrometry.
Achieve superior selectivity and enable more proteins to be identified while reducing time-consuming sample preparation steps with the next generation differential ion mobility Thermo Scientific™ FAIMS Pro™ interface.
The FAIMS Pro interface reduces chemical noise and matrix interferences, resulting in improved assay robustness and increased sensitivity. The FAIMS Pro interface offers separation based on a combination of factors, such as charge state, shape, conformation, and size of gas phase ions. It also improves dynamic range and peak capacity, which are particularly beneficial in some proteomics applications.
Add a new dimension of selectivity to every LC/MS proteomic laboratory with the high-field asymmetric waveform ion mobility spectrometry (FAIMS) Pro interface.
Unsurpassed selectivity can be achieved when FAIMS Pro technology is used in combination with the high-resolution accurate mass of a Thermo Scientific™ Orbitrap™ Fusion™ Lumos™ Tribrid™ MS and Thermo Scientific™ Orbitrap™ Fusion™ Tribrid™ MS. FAIMS technology can also enhance MSn analyses on industry-leading Thermo Scientific™ ion trap LC-MS systems.
Other benefits of the FAIMS Pro interface include:
Compatible with