FirstDefender RMX Handheld Chemical Identification Analyzer is an all-optical Raman system designed for use by first responders, homeland security, military, law enforcement and forensic chemistry personnel.
Fast, Accurate Identification
Quickly identifies unknown solid and liquid chemicals.
Built for Field Use
MIL-STD-810G and IP67 tested and certified.
Improved Automatic Mixture Analysis
Sophisticated algorithms automatically determine presence of mixed and contaminated chemicals.
Point-and-Shoot Identification
Operates directly through sealed glass or plastic containers, avoiding exposure to potentially harmful substances.
Extensive Substance Library
FirstDefender RMX portable chemistry analyzer Identifies explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more.
Flexible use modes: handheld point-and-shoot with fixed probe, mount to select tactical robots, or integrated vial mode.
Menu driven user interface requires minimal training.
No scheduled maintenance or calibration.
Non-destructive testing maintains sample integrity.
Powered using rechargeable battery; wall plug; or 123a disposable batteries.
Fully operable over a wide temperature range -4° to +122°F (-20° to +50 °C).
Integrated NIOSH and CAMEO database provides detailed hazard data on scene.
24/7 technical and reachback support provided for all units.