FastDigest XbaI
Inquire about OEM or Commercial Supply version of this product here.
FastDigest XbaI
Thermo Scientific™

FastDigest XbaI

5' T ↓C T A G A 3' 3' A G A T C ↑T 5' Thermo Scientific FastDigest XbaIRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FD0685750 μL (750 reactions)
FD0684300 μL (300 reactions)
Catalog number FD0685
Price (USD)/ 750 reactions
112.65
Online exclusive
130.00 
Save 17.35 (13%)
-
Add to cart
Quantity:
750 μL (750 reactions)
Request bulk or custom format
Price (USD)/ 750 reactions
112.65
Online exclusive
130.00 
Save 17.35 (13%)
Add to cart
FastDigest XbaI
Catalog numberFD0685
Price (USD)/ 750 reactions
112.65
Online exclusive
130.00 
Save 17.35 (13%)
-
Add to cart
5'  T ↓C  T  A  G  A   3' 
3'  A  G  A  T  C ↑T   5' 

Thermo Scientific FastDigest XbaI restriction enzyme recognizes T^CTAGA site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer.

Thermo Scientific FastDigest XbaI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features

• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Methylation SensitivityDam Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive
EnzymeXba I
Compatible Buffer10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat InactivationYes
Optimal Reaction Temperature37°C
Type IIS RENo
Quantity750 μL (750 reactions)
Product TypeRestriction Enzyme
Research CategoryTraditional Cloning
Product LineFastDigest™
Overhang5' protruding ends
Inactivation Temperature65°C
Unit Size750 reactions
Contents & Storage
• FastDigest XbaI (750 μL)
• 10X FastDigest Buffer (2 x 1.0 mL)
• 10X FastDigest Green Buffer (2 x 1.0 mL)

Store at –20°C.