FastDigest SmaI
Inquire about OEM or Commercial Supply version of this product here.
FastDigest SmaI
Thermo Scientific™
FastDigest SmaI
5' CCC ↓GGG 3'3' GGG ↑CCC 5'Thermo Scientific FastDigest SmaI is one of an advanced line of fast restriction enzymesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FD0664200 μL (200 Reactions)
FD0663100 μL (100 Reactions)
Catalog number FD0664
Price (USD)/ 200 reactions
63.65
Online exclusive
72.00 
Save 8.35 (12%)
-
Add to cart
Quantity:
200 μL (200 Reactions)
Request bulk or custom format
Price (USD)/ 200 reactions
63.65
Online exclusive
72.00 
Save 8.35 (12%)
Add to cart
FastDigest SmaI
Catalog numberFD0664
Price (USD)/ 200 reactions
63.65
Online exclusive
72.00 
Save 8.35 (12%)
-
Add to cart
5' CCC ↓GGG 3'
3' GGG ↑CCC 5'
Thermo Scientific FastDigest SmaI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers. The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps.
This restriction enzyme recognizes CCC^GGG site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer. Isoschizomers: TspMI, XmaCI, XmaI.
DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Features
  • 100% activity of all FastDigest enzymes in the universal buffer
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5–15 minutes
  • Direct loading on gels
  • No star activity
  • 176 FastDigest enzymes available
Applications
  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP analysis
Notes
  • The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer.
  • For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.
  • For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Methylation SensitivityCpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
EnzymeSma I
Compatible Buffer10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat InactivationYes
Optimal Reaction Temperature37°C
Type IIS RENo
Quantity200 μL (200 Reactions)
Product TypeRestriction Enzyme
Research CategoryTraditional Cloning
Product LineFastDigest™
OverhangBlunt
Inactivation Temperature65°C
Unit Size200 reactions
Contents & Storage
FastDigest SmaI (200 μL), 10X FastDigest Buffer (1.0 mL), 10X FastDigest Green Buffer (1.0 mL), Store at –20°C.