FastDigest Eco31I (IIs class)
Inquire about OEM or Commercial Supply version of this product here.
FastDigest Eco31I (IIs class)
Thermo Scientific™

FastDigest Eco31I (IIs class)

5' G G T C T C N1↓ 3'3' C C A G A G N5↑ 5'Thermo Scientific FastDigest Eco31IRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FD0294100 μL (100 Reactions)
FD029350 μL (50 Reactions)
Catalog number FD0294
Price (USD)/ 100 reactions
68.65
Online exclusive
75.75 
Save 7.10 (9%)
-
Add to cart
Quantity:
100 μL (100 Reactions)
Request bulk or custom format
Price (USD)/ 100 reactions
68.65
Online exclusive
75.75 
Save 7.10 (9%)
Add to cart
FastDigest Eco31I (IIs class)
Catalog numberFD0294
Price (USD)/ 100 reactions
68.65
Online exclusive
75.75 
Save 7.10 (9%)
-
Add to cart
5'  G  G  T  C  T  C  N1 3'
3'  C  C  A  G  A  G  N5 5'

Thermo Scientific FastDigest Eco31I (IIs class) restriction enzyme recognizes GGTCTC(1/5)^ site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer. Isoschizomers: BsaI, Bso31I, BspTNI.

Thermo Scientific FastDigest Eco31I is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features

• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Methylation SensitivityCpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
EnzymeEco31I
Compatible Buffer10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat InactivationYes
Optimal Reaction Temperature37°C
Type IIS REYes
Quantity100 μL (100 Reactions)
Product TypeRestriction Enzyme
Research CategoryScarless Cloning
Product LineFastDigest™
Overhang5' protruding ends
Inactivation Temperature65°C
Unit Size100 reactions
Contents & Storage
• FastDigest Eco31I (100 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.