Bovine Genotypes Panel 2.2 Kit
Product Image
Thermo Scientific™

Bovine Genotypes Panel 2.2 Kit

Thermo Scientific Bovine Genotypes Panel 2.2 Kit is offered for cattle parentage testing and identification using Short Tandem Repeat (STR)Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
F847S100 Reactions
F847L500 Reactions
Catalog number F847S
Price (USD)/ 100 reactions
684.00
-
Add to cart
No. of Reactions:
100 Reactions
Price (USD)/ 100 reactions
684.00
Add to cart
Bovine Genotypes Panel 2.2 Kit
Catalog numberF847S
Price (USD)/ 100 reactions
684.00
-
Add to cart
Thermo Scientific Bovine Genotypes Panel 2.2 Kit is offered for cattle parentage testing and identification using Short Tandem Repeat (STR) DNA loci.

The Bovine Genotypes Panel 2.2 allows co-amplification of the 6 microsatellites in a single multiplex PCR reaction. One primer from each primer pair is end-labeled with a fluorescent dye. After PCR, the fragments are separated and detected in a single electrophoresis injection, using an automated electrophoresis instrument (see Figure 2). In addition, the kit includes bovine control DNA for verification of acceptable PCR and electrophoresis conditions. Reagents and protocols of the Bovine Genotypes Panel 2.2 have been optimized to deliver similar peak sizes (amplification yields) for each loci. Allele callings obtained with this kit represent the true alleles of an individual, instead of 'plus-A peaks' or 'split peaks' typically interpreted when using e.g. a conventional Taq DNA polymerase. The kit employs Phusion Hot Start DNA Polymerase that has a proofreading activity and eliminates the 'plus-A peaks'.

Thermo Scientific Bovine Genotypes Panel 2.2 encompasses the following six STR loci: SPS113, RM067, CSRM60, MGTG4B, CSSM66, and ILSTS006. These are among the list of loci recommended by the International Society for Animal Genetics (ISAG) and the Food and Agriculture Organization of the United Nations (FAO) for genetic studies of domestic animals.

The Bovine Genotypes Panel 2.2 is intended to be used as a complement to the Bovine Genotypes Panel 1.2 when more STR loci are required for resolving a parentage case. The microsatellite alleles of the Bovine Genotypes Panel 2.2 kit can be separated and detected in the same single electrophoresis injection with the microsatellite alleles of the Bovine Genotypes Panel 1.2 kit (see Figure 2).

Highlights

• Various kit choices that include STR loci recommended by ISAG
• Single multiplex reaction - less pipetting steps
• Optimized reagents
• Control DNA provided for verification of PCR and electrophoresis conditions
• Allele calling represents true alleles (no plus-A peaks)

This product has been developed in collaboration with Maxxam Analytics Inc.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodCapillary Electrophoresis
For Use With (Application)PCR
No. of Reactions100 Reactions
Product TypeBovine Genotypes Panel 2.2 Kit
TargetBovine Genotyping and Parentage Testing
TechniqueSTR (Short Tandem Repeat) Analysis
Unit Size100 reactions
Contents & Storage
Contains:
• 1.1 mL Bovine Genotypes Master Mix
• 1.1 mL Bovine Genotypes Panel 2.2 Primer Mix
• 30 μL Bovine Genotypes Control DNA001

Store at -20°C. Primer Mix should be protected from light at all the times.