Phusion Hot Start II High-Fidelity PCR Master Mixes
Inquire about OEM or Commercial Supply version of this product here.
Phusion Hot Start II High-Fidelity PCR Master Mixes
Thermo Scientific™

Phusion Hot Start II High-Fidelity PCR Master Mixes

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Reactions
F565SColorless100 Reactions
F566SGreen100 Reactions
F566LGreen500 Reactions
F565LColorless500 Reactions
Catalog number F565S
Price (USD)/ 100 reactions
197.60
Special Offer
Online exclusive
Ends: 16-Dec-2024
247.00 
Save 49.40 (20%)
-
Add to cart
Color:
Colorless
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)/ 100 reactions
197.60
Special Offer
Online exclusive
Ends: 16-Dec-2024
247.00 
Save 49.40 (20%)
Add to cart
Phusion Hot Start II High-Fidelity PCR Master Mixes
Catalog numberF565S
Price (USD)/ 100 reactions
197.60
Special Offer
Online exclusive
Ends: 16-Dec-2024
247.00 
Save 49.40 (20%)
-
Add to cart
Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Highlights

  • Reaction set up at room temperature
  • No non-specific amplification and primer degradation during reaction set up
  • Zero-time reactivation due to unique hot start technology
  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15-30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gels

With Phusion Hot Start II High-Fidelity DNA Polymerases amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3'→5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.

Phusion Green Hot Start II High-Fidelity PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phusion Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. The buffer also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel.

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning
  • Mutagenesis
  • Microarray

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
GC-Rich PCR PerformanceMedium
PolymerasePhusion Hot Start II High-Fidelity DNA Polymerase
Reaction SpeedFast
Product TypeHot Start High-Fidelity PCR Master Mix
Quantity100 x 50 μL Reactions
Shipping ConditionDry Ice
For Use With (Application)Hot-start PCR, High-fidelity PCR
Concentration2X
Fidelity (vs. Taq)52X
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
OverhangBlunt
Reaction FormatSuperMix or Master Mix
Size (Final Product)20 kb or less
ColorColorless
Unit Size100 reactions
Contents & Storage
• 2X Phusion Hot Start II High-Fidelity PCR Master Mix
• Nuclease-free water
• DMSO

2X Phusion Hot Start II High-Fidelity PCR Master Mix provides 1.5 mM MgCl2 in the final 1X concentration.
Store at –20°C.