Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca
2+. Fluo-5F, fluo-5N, and fluo-4ff are analogs of fluo-4 with lower Ca
2+-binding affinity, making them suitable for detecting intracellular calcium levels in the 1 μM to 1 mM range that would saturate the response of fluo-3 and fluo-4. Cells may be physically loaded with the cell-impermeant salt forms of these indicators using patch pipette or microinjection or our
Influx™ pinocytotic cell-loading reagent. These indicators are compatible with excitation at 488 nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications.
Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›Calcium Indicator (Cell-Impermeant Salts) Specifications:
• Label (Ex/Em of Ca2+–bound form): Fluo-4FF (494/516 nm)
• Fluorescence intensity increase upon binding Ca2+: >100 fold
• Kd for Ca2+ in buffer: ∼9.7 μM
• Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelengthUsing TPEN to Control Heavy Metal Cations In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn
2+, Zn
2+, Pb
2+) with substantially higher affinity than Ca
2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator
TPEN.
More Choices for Fluorescent Calcium Indicators We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review
Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.
For UV-excitable Ca
2+ indicators, protein-based Ca
2+ indicators, conjugates of Ca
2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg
2+, Zn
2+) review
Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.