EASY-Spray™ PepMap™ Neo UHPLC Columns
EASY-Spray™ PepMap™ Neo UHPLC Columns
Thermo Scientific™

EASY-Spray™ PepMap™ Neo UHPLC Columns

Separate challenging peptide mapping samples with Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
ES75150PN150 mm
ES75500PN500 mm
ES75750PN750 mm
Catalog number ES75150PN
Price (USD)/ Each
1,415.96
Add to cart
Length (Metric):
150 mm
Price (USD)/ Each
1,415.96
Add to cart
EASY-Spray™ PepMap™ Neo UHPLC Columns
Catalog numberES75150PN
Price (USD)/ Each
1,415.96
-
Add to cart
Separate challenging peptide mapping samples using Thermo Scientific™ EASY-Spray™ PepMap™ Neo UHPLC columns. These columns feature easy connectivity, high reproducibilty, and excellent separations. Our Neo columns provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to obtain greater sample coverage and sample insights. The Click-and-Spray format of EASY-Spray columns enables perfect connections to our mass spectrometers every time.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — EASY-Spray™ columns click into position with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
Specifications
Column FormatAnalytical Column, EASY-Spray™
Diameter (Metric)75 μm
Length (Metric)150 mm
Max. Pressure22,000 psi (1500 bar)
Particle Size2 μm
pH2 to 8
Pore Size100 Å
Surface Area450 m2/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineEasy Spray
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load15%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
DescriptionEasy-Spray™ PepMap™ Neo 2 μm C18 75 μm X 150 mm
For Use With (Application)Bottom-up Proteomics, LC-MS
Unit SizeEach