PvuI (10 U/μL)
Inquire about OEM or Commercial Supply version of this product here.
PvuI (10 U/μL)
Thermo Scientific™

PvuI (10 U/μL)

5' C G A T ↓C G 3' 3' G C ↑T A G C 5' Thermo Scientific PvuI restrictionRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
ER0621300 U
ER06221500 U
Catalog number ER0621
Price (USD)/ 300 units
100.65
Online exclusive
113.00 
Save 12.35 (11%)
-
Add to cart
Quantity:
300 U
Request bulk or custom format
Price (USD)/ 300 units
100.65
Online exclusive
113.00 
Save 12.35 (11%)
Add to cart
PvuI (10 U/μL)
Catalog numberER0621
Price (USD)/ 300 units
100.65
Online exclusive
113.00 
Save 12.35 (11%)
-
Add to cart
5'  C  G  A  T ↓C  G   3' 
3'  G  C ↑T  A  G  C   5' 

Thermo Scientific PvuI restriction enzyme recognizes CGAT^CG sites and cuts best at 37°C in R buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion. Isoschizomers: BpvUI, MvrI, Ple19I.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 U/μL
Methylation SensitivityCpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
EnzymePvu I
Compatible Buffer10X Buffer R
Sensitive to Heat InactivationYes
Optimal Reaction Temperature37°C
Type IIS RENo
Quantity300 U
Product TypeRestriction Enzyme
Research CategoryTraditional Cloning
Unit Size300 units