DNA Polymerase I (10 U/μL)
Inquire about OEM or Commercial Supply version of this product here.
DNA Polymerase I (10 U/μL)
Thermo Scientific™

DNA Polymerase I (10 U/μL)

Thermo Scientific DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonucleaseRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EP00422500 U
EP0041500 U
Catalog number EP0042
Price (USD)/ 2,500 units
334.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
394.00 
Save 59.10 (15%)
-
Add to cart
Quantity:
2500 U
Request bulk or custom format
Price (USD)/ 2,500 units
334.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
394.00 
Save 59.10 (15%)
Add to cart
DNA Polymerase I (10 U/μL)
Catalog numberEP0042
Price (USD)/ 2,500 units
334.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
394.00 
Save 59.10 (15%)
-
Add to cart
Thermo Scientific DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.

Highlights

• Incorporates modified nucleotides (e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
• Active in multiple buffers, including restriction enzyme, PCR, and RT buffers

Applications

• DNA labeling by nick-translation in conjunction with DNase
• Second-strand synthesis of cDNA in conjunction with RNase H
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionDNA Polymerase I
PolymeraseDNA Polymerase I
Quantity2500 U
Concentration10 U/μL
Unit Size2,500 units