RiboLock RNase Inhibitor (40 U/μL)
GMP-quality RNase inhibitor is available for RNA vaccine and therapeutic development. See details
Inquire about OEM or Commercial Supply version of this product here.
RiboLock RNase Inhibitor (40 U/μL)
Thermo Scientific™

RiboLock RNase Inhibitor (40 U/μL)

Thermo Scientific RiboLock RNase Inhibitor inhibits the activity of RNases A,B and C by binding them in a noncompetitive modeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EO038424 x 2500 U
EO03812500 U
EO03824 x 2500 U
Catalog number EO0384
Price (USD)/ 24 x 2,500 units
1,287.65
Online exclusive
1472.00 
Save 184.35 (13%)
-
Add to cart
Quantity:
24 x 2500 U
Request bulk or custom format
Price (USD)/ 24 x 2,500 units
1,287.65
Online exclusive
1472.00 
Save 184.35 (13%)
Add to cart
RiboLock RNase Inhibitor (40 U/μL)
Catalog numberEO0384
Price (USD)/ 24 x 2,500 units
1,287.65
Online exclusive
1472.00 
Save 184.35 (13%)
-
Add to cart
Thermo Scientific RiboLock RNase Inhibitor inhibits the activity of RNases A,B and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.

Highlights

Performs under a wide range of reaction conditions
Protects RNA from degradation at temperatures up to 55°C

Note

DTT provided in the Storage Buffer ensures stability during long term storage, but is not necessary for inhibitor activity. Recommended concentration 1 U/ μL of a reaction mixture.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration40 U/μL
Compatible BufferStorage Buffer
Quantity24 x 2500 U
Product TypeRNase Inhibitor
Product LineRiboLock™
Unit Size24 x 2,500 units