T4 Polynucleotide Kinase (10 U/μL)
Inquire about OEM or Commercial Supply version of this product here.
T4 Polynucleotide Kinase (10 U/μL)
Thermo Scientific™

T4 Polynucleotide Kinase (10 U/μL)

Thermo Scientific™ T4 Polynucleotide Kinase (T4 PNK) catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EK00322500 U
EK0031500 U
Catalog number EK0032
Price (USD)/ 2,500 units
221.00
Special Offer
Online exclusive
Ends: 16-Dec-2024
260.00 
Save 39.00 (15%)
-
Add to cart
Quantity:
2500 U
Request bulk or custom format
Price (USD)/ 2,500 units
221.00
Special Offer
Online exclusive
Ends: 16-Dec-2024
260.00 
Save 39.00 (15%)
Add to cart
T4 Polynucleotide Kinase (10 U/μL)
Catalog numberEK0032
Price (USD)/ 2,500 units
221.00
Special Offer
Online exclusive
Ends: 16-Dec-2024
260.00 
Save 39.00 (15%)
-
Add to cart
Thermo Scientific™ T4 Polynucleotide Kinase (T4 PNK) catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group of single- and double-stranded DNAs and RNAs, oligonucleotides, or nucleoside 3'-monophosphates (forward reaction). The reaction is reversible. In the presence of ADP, T4 Polynucleotide Kinase exhibits 5'-phosphatase activity and catalyzes the exchange of phosphate groups between 5'-P-oligo-polynucleotides and ATP (exchange reaction). The enzyme is also a 3'-phosphatase.

Highlights
• Active in Thermo Scientific restriction enzyme, RT, and T4 DNA ligase buffers

Applications
• Labeling 5' -termini of nucleic acids to be used as:
    --Probes for hybridization
    --Probes for transcript mapping
    --Markers for gel electrophoresis
    --Primers for DNA sequencing
    --Primers for PCR
• 5'-phosphorylation of oligonucleotides, PCR products, other DNA or RNA prior to ligation
• Phosphorylation of PCR primers
• Detection of DNA modification by the [32P]-postlabeling assay
• Removal of 3'-phosphate groups

Notes
• The 5'-termini of nucleic acids can be labeled by either the forward or the exchange reaction.
• Polyethylene glycol (PEG) and spermidine improve the rate and efficiency of the phosphorylation reaction. PEG is used in the exchange reaction mixture.
• Since T4 Polynucleotide Kinase is inhibited by ammonium ions, use sodium acetate to precipitate DNA prior to phosphorylation.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 U/μL
EnzymeT4 Polynucleotide Kinase
Compatible Buffer10X Reaction Buffer
Quantity2500 U
Product TypeT4 Polynucleotide Kinase
Unit Size2,500 units