DH5α Competent Cells
Inquire about OEM or Commercial Supply version of this product here.
DH5α Competent Cells
Thermo Scientific™

DH5α Competent Cells

Thermo Scientific DH5α Competent Cells are high efficiency, chemically competent E. coli cells ideal for construction of gene banks orRead more
Have Questions?
Catalog number EC0112
Price (USD)/ 10 x 100 µL
137.65
Online exclusive
154.00 
Save 16.35 (11%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 10 x 100 µL
137.65
Online exclusive
154.00 
Save 16.35 (11%)
Add to cart
DH5α Competent Cells
Catalog numberEC0112
Price (USD)/ 10 x 100 µL
137.65
Online exclusive
154.00 
Save 16.35 (11%)
-
Add to cart
Thermo Scientific DH5α Competent Cells are high efficiency, chemically competent E. coli cells ideal for construction of gene banks or generation of cDNA libraries using plasmid-derived vectors. The φ80dlacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors to allow blue/white colony screening on bacterial agar plates containing Bluo-Gal or X-Gal. RecA1 and endA1 mutations in DH5α cells improve insert stability and quality of the extracted plasmid DNA.

• High transformation efficiency: >1x109 cfu/μg pUC19 DNA
• Suitable for routine and high-throughput cloning applications
• Genetic markers that allow for blue/white colony screening
• Convenient two reactions per vial packaging
• S.O.C. Outgrowth medium included

Applications
DH5α Competent Cells are suitable for variety of applications requiring high transformation efficiency:
• Efficient DNA cloning derived from PCR, cDNA-generation reactions
• recA1 marker facilitates working with difficult to transform DNA
• Ideal for generation of cDNA libraries using plasmid-derived vectors
• Site-directed mutagenesis

Genotype
F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeCompetent Cell
Contains F' EpisomeLacks F' Episome
EfficiencyHigh Efficiency (>1x10^9 cfu/μg pUC19 DNA)
Cell TypeChemically Competent
LibrariescDNA
Cloning Methylated DNANo
Bacterial or Yeast StrainDH5α
Blue/White ScreeningYes
High-throughput CompatibilityNot High-throughput Compatible
Propagating ccdB VectorsNot for ccdB vector propagation
Shipping ConditionDry Ice
SpeciesE. coli
FormatTube
Product LineThermo Scientific™
For Use With (Application)Cloning
Quantity10 x 100 μL
Unit Size10 x 100 µL
Contents & Storage
• 10 x DH5α Competent Cells (100 μL)
• pUC19 DNA (50 μL) (10 pg/μL)
• S.O.C. Medium (5 mL)

Store at -80°C.