EnzChek™ Protease Assay Kit, red fluorescence, 100-1000 assays
The EnzChek™ Protease Assay Kit, red fluorescence, is a fast, simple, and direct fluorescence-based assay for detecting metallo-, serine, acid,Read more
Have Questions?
Catalog number E6639
Price (USD)/ 1 kit
499.00
-
Add to cart
Price (USD)/ 1 kit
499.00
Add to cart
EnzChek™ Protease Assay Kit, red fluorescence, 100-1000 assays
Catalog numberE6639
Price (USD)/ 1 kit
499.00
-
Add to cart
The EnzChek™ Protease Assay Kit, red fluorescence, is a fast, simple, and direct fluorescence-based assay for detecting metallo-, serine, acid, and sulfhydryl proteases. The accompanying increase in fluorescence, which can be measured with a fluorescence microplate reader, is proportional to protease activity.
• Generalized mechanism allows for measurement of a variety of proteases • Can be used to continuously measure protease kinetics • Excitation/emission similar to Texas Red™ dye (589/617 nm)
This EnzChek™ Protease Assay Kit contains a casein derivative that has been extensively labeled with the red-fluorescent BODIPY™ TR-X dye, which results in a quenching of the fluorescent dye. Protease-catalyzed hydrolysis releases the highly-fluorescent BODIPY™ TR-X dye-labeled peptides, allowing for quantitative detection of protease activity in solution. The red-fluorescent BODIPY™ TR-X dye has excitation and emission spectra similar to those of the Texas Red™ fluorophore.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence Intensity
For Use With (Application)Protease Assay
For Use With (Equipment)Fluorescence Microplate Reader
Product TypeProtease Assay
Product LineEnzChek™
Quantity1 kit
Shipping ConditionRoom Temperature
Substrate PropertiesProtein-Based Substrate
Substrate TypeProtease Substrate
Target EnzymeProteases
Unit Size1 kit
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.