Catalog Number | Description |
---|---|
E40966 | SuperBoost™ EverBlue HRP Kit, goat anti-mouse IgG |
E40964 | SuperBoost™ EverBlue HRP Substrate Kit |
E40972 | SuperBoost™ EverBlue HRP Substrate 5X Kit |
E40968 | SuperBoost™ EverBlue HRP Kit, goat anti-rabbit IgG |
E40970 | SuperBoost™ EverBlue HRP Kit, Streptavidin |
E40963 | SuperBoost™ EverRed HRP Substrate Kit |
E40971 | SuperBoost™ EverRed HRP Substrate 5X Kit |
E40965 | SuperBoost™ EverRed HRP Kit, goat anti-mouse IgG |
E40967 | SuperBoost™ EverRed HRP Kit, goat anti-rabbit IgG |
E40969 | SuperBoost™ EverRed HRP Kit, Streptavidin |
Tyramide signal deposition in the Tyramide SuperBoost™ kits uses the catalytic activity of horseradish peroxidase (HRP) for high-density labeling of a target protein, enabling permanent colorimetric tissue staining with SuperBoost EverRed and/or EverBlue that is wash resistant, water/organic solvent mountable, and archivable. Unlike many other colorimetric IHC staining methods, SuperBoost EverRed and EverBlue staining is covalently attached to the target resulting in a sharp and intense chromogenic signal that is maintained without needing to air-dry tissue or use unconventional dehydration/clearing protocols.
The kits are simple to use and easily adapted to standard IHC experimental protocols using any tissue type, including formalin-fixed, paraffin-embedded (FFPE) and cryopreserved tissue. Tissues labeled using the SuperBoost EverRed and EverBlue kits can be imaged using any type of brightfield microscope, producing high-resolution images.
Features of the SuperBoost EverRed and EverBlue HRP Kits include:
• Permanent colorimetric staining that is also fluorescent, allowing confirmation of the staining pattern with both brightfield and fluorescence imaging
• Compatibility with other common colorimetric counterstains
• Multiplexability between EverRed, EverBlue, and DAB
How does SuperBoost color signal anchoring work?
SuperBoost EverRed and EverBlue HRP Kits use a simple workflow similar to standard IHC staining methods. For SuperBoost EverRed and EverBlue HRP tissue staining, an antigen is detected by a primary antibody, followed by staining with a secondary antibody or streptavidin (for biotinylated primary antibodies) conjugated with poly-HRP. HRP then enzymatically activates a chromogen tyramide, resulting in localized covalent deposition of the activated tyramide derivative at tyrosine residues on and surrounding the protein epitope targeted by the primary antibody. Treatment of the labeled tissue with the EverRed or EverBlue reagent then results in a permanent colorimetric signal at the site of the covalent tyramide deposition.
SuperBoost EverRed/EverBlue HRP Kits, goat anti-mouse IgG/goat anti-rabbit
The SuperBoost EverRed/EverBlue HRP Kits, goat anti-mouse IgG/goat anti-rabbit can be used for permanent red or blue colorimetric detection of mouse or rabbit IgG primary antibodies in IHC applications. The kits include the goat anti-mouse IgG or goat anti-rabbit IgG poly horseradish peroxidase (HRP) conjugate, tyramide HRP substrate, and SuperBoost EverRed or EverBlue reagent along with the buffers and reagents needed for tissue staining and color development. Sufficient reagents are provided to stain 150 tissue sections.
The four options for these kits include:
• SuperBoost™ EverRed HRP Kit, goat anti-mouse IgG
• SuperBoost™ EverRed HRP Kit, goat anti-rabbit IgG
• SuperBoost™ EverBlue HRP Kit, goat anti-mouse IgG
• SuperBoost™ EverBlue HRP Kit, goat anti-rabbit IgG
SuperBoost EverRed/EverBlue HRP Kits, Streptavidin
The SuperBoost EverRed/EverBlue HRP Kits, Streptavidin can be used for permanent red or blue colorimetric detection of biotinylated primary antibodies in IHC applications. The kits include the streptavidin HRP conjugate, tyramide HRP substrate, and SuperBoost EverRed or EverBlue reagent along with the buffers and reagents needed for tissue staining and color development. Sufficient reagents are provided to stain 150 tissue sections.
SuperBoost EverRed/EverBlue HRP Substrate Kits
The SuperBoost EverRed/EverBlue HRP Substrate Kits include the tyramide HRP substrate and SuperBoost EverRed or EverBlue reagent for permanent red or blue colorimetric detection in IHC applications using your own HRP conjugate. Sufficient reagents are provided to stain 150 tissue sections or 5 x 150 tissue sections.