Epidermal Growth Factor, Biotinylated, complexed to Texas Red™ Streptavidin (Texas Red™ EGF complex)
Epidermal Growth Factor, Biotinylated, complexed to Texas Red™ Streptavidin (Texas Red™ EGF complex)
Invitrogen™
Epidermal Growth Factor, Biotinylated, complexed to Texas Red™ Streptavidin (Texas Red™ EGF complex)
We offer several conjugates of epidermal growth factor that are useful for the detection of EGF receptor in cells. FluorescentlyRead more
Have Questions?
Catalog number E3480
also known as E-3480
Price (USD)/ 100 µg
530.00
-
Add to cart
Price (USD)/ 100 µg
530.00
Add to cart
Epidermal Growth Factor, Biotinylated, complexed to Texas Red™ Streptavidin (Texas Red™ EGF complex)
Catalog numberE3480
Price (USD)/ 100 µg
530.00
-
Add to cart
We offer several conjugates of epidermal growth factor that are useful for the detection of EGF receptor in cells. Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more. The conjugates incorporating fluorescein (FITC), Oregon Green™ 514, and tetramethylrhodamine were constructed with the dye directly attached to EGF. The conjugates of EGF with Alexa Fluor™ dyes and Texas Red™ dye are complexes of the dye–streptavidin molecule and biotinylated EGF. The biotinylated conjugates utilize biotin-XX, which contains a long spacer arm designed to enhance the probe’s affinity for the EGF receptor.
Epidermal Growth Factor Conjugate Specifications:
• EGF molecule: 53 amino acids, MW=6,045 Da
• Label (Ex/Em): Biotin-XX complexed with Texas Red™ streptavidin (∼595/615 nm)
• Number of fluorophore molecules on each EGF molecule: ∼2–3
• Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter
Find More Receptor Binding and Phagocytosis Probes
We offer a number of fluorescently labeled probes for studying receptor-mediated endocytosis, membrane markers for endocytosis and exocytosis, and methods for detecting internalized fluorescent ligands. Review Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes™ Handbook for more information on these products.
For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Detection MethodFluorescence
Product TypeGrowth Factor
Label or DyeClassic Dyes
Product LineTexas Red™
Quantity100 μg
Shipping ConditionWet Ice
Unit Size100 µg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.