The EnzChek™ Lysozyme Assay Kit provides researchers with a simple and sensitive assay to measure lysozyme activity in solution. TheRead more
Have Questions?
Catalog number E22013
Price (USD)/ 400 assays
418.65
Online exclusive
422.00
Save 3.35
-
Add to cart
Price (USD)/ 400 assays
418.65
Online exclusive
422.00
Save 3.35
Add to cart
EnzChek™ Lysozyme Assay Kit
Catalog numberE22013
Price (USD)/ 400 assays
418.65
Online exclusive
422.00
Save 3.35
-
Add to cart
The EnzChek™ Lysozyme Assay Kit provides researchers with a simple and sensitive assay to measure lysozyme activity in solution. The increase in fluorescence resulting from lysozyme activity is measured using a fluorometer or fluorescence microplate reader.
• Detect lysozyme activity concentrations as low as 20 U/mL • Use standard fluorescein (FITC) excitation/emission settings • Format allows for continuous measurement of lysozyme activity
The assay utilizes an innovative approach to measure lysozyme activity. Micrococcus lysodeikticus cell walls are used that have been extensively labeled with fluorescein causing quench of the fluorescence signal. Active lysozyme enzyme hydrolyzes the b-(1-4)-glucosidic linkages between the N-acetylmuramic acid and N-acetyl-D-glucosamine residues in the mucopolysaccharide cell wall, relieving the quenching and yielding a dramatic increase in fluorescence that is proportional to lysozyme activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence Intensity
For Use With (Application)Lysozyme Assay
For Use With (Equipment)Fluorescence Microplate Reader
Product TypeLysozyme Assay
Product LineEnzChek™
Quantity400 assays
Shipping ConditionRoom Temperature
Substrate PropertiesLipid-Based Substrate
Substrate TypeLysozyme Substrate
Target EnzymeLysozyme
Unit Size400 assays
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.