Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns
Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns
Actual product may vary
Thermo Scientific™

Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns

Separate challenging peptide mapping samples with Thermo Scientific™ Double nanoViper™ PepMap™ Neo UHPLC columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Product Line
DNV75150PN150 mmDouble nanoViper
DNV75500PN500 mmDouble nanoViper
DNV75750PN750 mmDouble nanoViper
SNV150150PN150 mmSingle nanoViper
DNV150150PN150 mmDouble nanoViper
SNV75750PN750 mmSingle nanoViper
Catalog number DNV75150PN
Price (USD)/ Each
1,239.44
Add to cart
Length (Metric):
150 mm
Product Line:
Double nanoViper
Price (USD)/ Each
1,239.44
Add to cart
Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns
Catalog numberDNV75150PN
Price (USD)/ Each
1,239.44
-
Add to cart
Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — Double nanoViper™ columns connect with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
Specifications
Column FormatAnalytical Column, Double nanoViper™
Diameter (Metric)75 μm
Length (Metric)150 mm
Max. Pressure22,000 psi (1500 bar)
Particle Size2 μm
pH2 to 8
Pore Size100 Å
Surface Area450 m2/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineDouble nanoViper
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load15%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
DescriptionDouble nanoViper™ PepMap™ Neo 2 μm C18 75 μm X 150 mm
For Use With (Application)Peptide Mapping, LC-MS
Unit SizeEach