Boost walkaway time, reduce human error, increase throughput and improve sample processing uniformity. The Thermo Scientific™ DNA Purification System is an automation tool for bead based methods using magnetic particles. It automates all steps involved in the purification of DNA samples: binding of DNA to magnetic beads, washing of the bound material, elution from the solid phase and transfer to a final destination plate.
Includes:
Standard lab table (2.00 × 2.44m) ; F3 robot system (6 axes of motion) ; Servo Gripper with Microplate Fingers; Magnetic Bead Settling Stations (four) ; 120 Plate Carousel; Microplate Feeding Stations (120-plate capacity); Dispenser with bead dispensing module; Plate Delidder with barcode reader; Microplate Shaker with pneumatic lock for reliable plate placement and retrieval (four stations); Biotek EL 405 Plate Washer (96-well with bead settling plate); Robbins Hydra-96™ Pipettor with bead settling plate; Polara™ software and instrument interfaces; Pentium™ III PC (or higher), 17in. Monitor, Windows™ NT
Ordering Alerts:
Optional Components