DeviceLink Connect Remote Monitoring System (Kits)
DeviceLink Connect Remote Monitoring System (Kits)
Actual product may vary
Thermo Scientific™

DeviceLink Connect Remote Monitoring System (Kits)

The DeviceLink Connect is a remote monitoring and asset management solution for laboratory equipment enabling knowledge based decisions by monitoring critical equipment parameters while providing data insights and real-time system notifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
DLCKTULTS101DeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, serial to USB ULT connection cable, cable mounting ties, and first year DeviceLink Connect subscription
DLCKTCO2I101DeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, serial to USB incubator connection cable, cable mounting ties, and first year DeviceLink Connect subscription
DLCKTCO2I102DeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, USB to USB incubator connection cable, cable mounting ties, and first year DeviceLink Connect subscription
DLCKTLRFZ101DeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, serial to USB refrigerator/freezer connection cable, cable mounting ties, and first year DeviceLink Connect subscription
DLCKTMOTE001DeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, cable mounting ties, and first year DeviceLink Connect subscription
Catalog number DLCKTULTS101
Price (USD)/ Each
-
Request A Quote
Description:
DeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, serial to USB ULT connection cable, cable mounting ties, and first year DeviceLink Connect subscription
Thermo Scientific™ DeviceLink Connect measures critical reliability parameters and uses advanced analytics to provide real-time monitoring and reporting for lab instruments. The system supports independent sensors to monitor your instrument, but it can also be connected via serial port directly to the instrument harnessing its on-board telemetry data, alarms, and events. All information is transmitted to the Thermo Connect cloud where you have 24/7 access to the information via web and mobile based platforms.

The DeviceLink Connect is a remote monitoring system that uses direct serial/USB based instrument connections and/or independent sensors to gather useful information about the operation of the equipment. Sending this data back to Thermo Connect cloud allows for 24/7 monitoring from anywhere in the world.

How do I choose the right kit?

If you have a piece of equipment that is located on the compatibility list, you would purchase the applicable kit for serial connection from the table above.

If you have a piece of equipment that is not on the compatibility list and you are using independent sensors, you will need to purchase the DLC Basic Mote Kit (DLCKTMOTE001), and whatever independent sensors you may want from the sensor's product family page.

How it works:

  1. Connect your DeviceLink Connect system
    • Supply power via the provided AC power cable
    • Connect your instrument via serial/USB cable to the DeviceLink Connect mote
    • Install (optional) independent sensors
  2. Configure your setup via the web-based Commissioner Application
  3. Establish a Thermo Connect cloud account
  4. Download the free mobile InstrumentConnect mobile application
  5. Monitor your equipment 24/7 from anywhere in the world

Network:

  • Wi-Fi: WPA Enterprise (802.1x) and WPA2-PSK protocols
  • Ethernet (LAN) connections also supported
  • Up to 7 days internal data storage during a network outage

Power and Battery:

  • Power-over-Ethernet (PoE) provides main power to the device.
  • PoE wall adapter provide or a PoE 802.3af standard connection can be used
  • Contains a rechargeable Li-ion backup battery providing up to 3 hrs. of backup power during continuous operation (2600 mAH, 3.6V)

Advanced InSight Analytics:

The DeviceLink Connect system is also compatible with Thermo Scientific™ InSight™ analytics, powered by KLATU™ which are designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:

  • Cabinet temperature
  • Electrical use and energy consumption
  • Door openings
  • Ambient temperature

When you purchase an InSight monitoring license and subscription (sold separately) your DeviceLink Connect data run through a number of algorithms utilizing the InSight software. With InSight analytics, you can determine and compare the assets' performance against industry performance standards adding that next level of asset management.

Installation and Services:

  • Customer (self) installation
  • Install services are sold separately if desired
  • Calibration sensors also available.

Available only in North America.

Specifications
Alarm TypesWeb/Mobile
Battery TypeLi-ion Back
Battery LifeUp to 3 Hr. Continuous Operation
DescriptionDeviceLink Connect Remote Monitoring System Kit includes: DeviceLink Connect mote, PoE power supply, USB commissioning dongle, serial to USB ULT connection cable, cable mounting ties, and first year DeviceLink Connect subscription
InterfaceEthernet (LAN) or Wi-Fi connections
Sensor TypesSerial to equipment, Temperature, Humidity, Current, CO2, Door Switch
For Use With (Application)Wireless Monitoring, Asset Management Solution
For Use With (Equipment)Thermo Scientific ULT freezers (via serial connection), All lab equipment (via independent sensors)
Electrical RequirementsPoE power cable and AC wall adapter (provided)
Unit SizeEach