Simplify dioxin and di-PCBs analysis in food with the sensitivity, efficient data processing and reporting, EU compliance, robustness, and increased productivity of the Thermo Scientific Dioxin Analyzer.
Ensure robust and sensitive quantitation of polychlorinated dibenzo-p-dioxins (PCDDs), polychlorinated dibenzo furans (PCDFs) and dioxin-like polychlorinated biphenyls (dl-PCBs) in food and feed samples with the Thermo Scientific™ Dioxin Analyzer. This workflow solution includes the Thermo Scientific™ TSQ™ 9000 Triple Quadrupole GC-MS/MS System equipped with an advanced ionization source (AEI) for enhanced sensitivity, and a range of productivity tools to deliver reliable and cost-effective routine analysis of dioxins and dl-PCBs in compliance with EU regulation 664/2017.
The Dioxin Analyzer includes the Thermo Scientific™ TSQ™ 9000 Triple Quadrupole GC-MS/MS System, consumables, system suitability check standard solutions, e-workflow with acquisition method, data processing software, isotope dilution calculation, and reporting methods in compliance with EU regulations. Comprehensive user guidelines for fast implementation and enablement of optimum performance are also included.
The Dioxin Analyzer offers the following benefits:
The TraceGOLD™ TG-Dioxin GC Columns (60m × 0.25mm, 0.25μm) enable isomer specificity for 2,3,7,8-TCDD and 2,3,7,8-TCDF. The columns feature the following benefits: